Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVRL2 Mouse anti-Human, Clone:4A11B12, Abnova™

Catalog No. 89334295 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-295 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-295 Supplier Abnova Corporation Supplier No. MAB17975
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human PVRL2.

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Specifications

Antigen PVRL2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4A11B12
Conjugate Unconjugated
Description poliovirus receptor-related 2 (herpesvirus entry mediator B)
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene PVRL2
Gene Alias CD112, HVEB, PRR2, PVRR2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 58-212 of human PVRL2.
Quantity 100 μg
Gene ID (Entrez) 5819
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.