Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PWAR1 Monoclonal Antibody (6A7H10)
GREENER_CHOICE

Catalog No. PIMA515445
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515445 Supplier Invitrogen™ Supplier No. MA515445
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15445 targets PAR1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15445 immunogen is purified recombinant fragment of PAR1 expressed in E. Coli.

PAR1: Prader-Willi/Angelman region-1 .
TRUSTED_SUSTAINABILITY

Specifications

Antigen PWAR1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6A7H10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene PWAR1
Gene Accession No. 0
Gene Alias D15S227E; PAR1; PAR-1; Prader Willi/Angelman region RNA 1; PWAR1
Gene Symbols PWAR1
Host Species Mouse
Immunogen Purified recombinant fragment of PAR1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145624
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.