Learn More
PWP1 Monoclonal Antibody (3A11)

Description
PWP1 Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | PWP1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3A11 |
| Concentration | 0.42 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PWP1 |
| Gene Accession No. | Q13610 |
| Gene Alias | 2310058A11Rik; 2610205J09Rik; AI098170; AL022879; AV083409; AW550690; C20orf152; endonuclein; IEF-SSP-9502; Keratinocyte protein IEF SSP 9502; nuclear phosphoprotein similar to S. cerevisiae PWP1; periodic tryptophan protein 1 homolog; Pwp1; PWP1 homolog (S. cerevisiae); PWP1 homolog, endonuclein; RGD1310705 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.