Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PWP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565221 Supplier Invitrogen™ Supplier No. PA565221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSLGKPAASLA VHTDKVQTLQ FHPFEAQTLI SGSYDKSVAL YDCRSPDESH RMWRFSGQIE RVTWNHFSPC HFLASTDDGF VYNLDARSDK P Highest antigen sequence identity to the following orthologs - mouse 87%, rat 89%.

The protein encoded by this gene contains several WD-40 repeats and is found mostly in the nucleus. The expression and localization of this protein are cell cycle dependent. Expression of this gene is upregulated in pancreatic adenocarcinoma. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PWP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PWP1
Gene Accession No. Q13610
Gene Alias 2310058A11Rik; 2610205J09Rik; AI098170; AL022879; AV083409; AW550690; C20orf152; endonuclein; IEF-SSP-9502; Keratinocyte protein IEF SSP 9502; nuclear phosphoprotein similar to S. cerevisiae PWP1; periodic tryptophan protein 1 homolog; Pwp1; PWP1 homolog (S. cerevisiae); PWP1 homolog, endonuclein; RGD1310705
Gene Symbols PWP1
Host Species Rabbit
Immunogen Recombinant Human PWP1. Recombinant protein control fragment (Product #RP-104694).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.