Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PWP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542781 Supplier Invitrogen™ Supplier No. PA542781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VQTGSIEGRH DLKTGRKELD KITAKHAAKG KAFTALCYSA DGHSILAGGM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

PWP2 belongs to the WD repeat PWP2 family. It contains 14 WD repeats. The exact function of PWP2 is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PWP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PWP2
Gene Accession No. Q15269
Gene Alias 6530411D08Rik; EHOC-17; periodic tryptophan protein 2 homolog; periodic tryptophan protein 2 homolog; LOW QUALITY PROTEIN: periodic tryptophan protein 2 homolog; Pwp2; PWP2 periodic tryptophan protein homolog; PWP2 periodic tryptophan protein homolog (yeast); PWP2 small subunit processome component; PWP2, small subunit processome component; PWP2, UTPB SSU processome subunit; PWP2H; UTP1; wdp103; WD-repeat protein p103
Gene Symbols PWP2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PWP2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5822
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.