Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PWWP2A Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$501.50

Specifications

Antigen PWWP2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170642
SDP
View Documents
Novus Biologicals
NBP170642
100 μL
Each of 1 for $501.50
Only null left
Add to Cart
 
Description

Description

PWWP2A Polyclonal specifically detects PWWP2A in Human samples. It is validated for Western Blot.
Specifications

Specifications

PWWP2A
Polyclonal
Rabbit
KIAA1935, MST101, MSTP101, PWWP domain containing 2A, PWWP domain-containing protein 2A
PWWP2A
IgG
Western Blot
Unconjugated
RUO
114825
Synthetic peptides corresponding to PWWP2A(PWWP domain containing 2A) The peptide sequence was selected from the C terminal of PWWP2A. Peptide sequence PQSRCTSTRSAGLNKWQLLHQTVTSPAAPLQCLTDHCGFRLGALKLTVKR.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.