Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PWWP2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561298 Supplier Invitrogen™ Supplier No. PA561298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSNTFQEGTE VKCEANGAVP DDPSPVPHPE LSLAESLWTS KPPPLFHEGA PYPPPLFIRD TYNQSIPQPP P Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

PWWP2A contains 1 PWWP domain. The exact function of PWWP2A remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PWWP2A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PWWP2A
Gene Accession No. Q96N64
Gene Alias 4631424J17Rik; AI891583; AU024105; D930040F23Rik; KIAA1935; MST101; MSTP101; PWWP domain containing 2A; PWWP domain-containing protein 2A; PWWP2A; RGD1305891
Gene Symbols PWWP2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PWWP2A. Recombinant protein control fragment (Product #RP-96930).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114825
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.