Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PWWP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140013
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VRVEQVVNGA LVVTVSCGER SFAGILLDCT KKSGLFGLPP LAPLPQVDES PVNDSHGRAP EEGDAEVMQL.
PWWP2B is a protein coding gene.
Specifications
PWWP2B | |
Polyclonal | |
Unconjugated | |
PWWP2B | |
bA432J24.1; pp8607; PWWP domain containing 2; PWWP domain containing 2B; PWWP domain-containing protein 2B; PWWP2; PWWP2B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170394 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NUJ5 | |
PWWP2B | |
Recombinant protein corresponding to Human PWWP2B. Recombinant protein control fragment (Product #RP-109668). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction