Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PWWP2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101797 Supplier Invitrogen™ Supplier No. PA5101797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PWWP2B.

PWWP2B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PWWP2B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PWWP2B
Gene Accession No. Q6NUJ5
Gene Alias bA432J24.1; pp8607; PWWP domain containing 2; PWWP domain containing 2B; PWWP domain-containing protein 2B; PWWP2; PWWP2B
Gene Symbols PWWP2B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PWWP2B(Accession Q6NUJ5), corresponding to amino acid residues V267-K317.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170394
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.