Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PX19 (PRELID1) Monoclonal Antibody (OTI2A6), TrueMAB™, OriGene
SDP

Catalog No. 502543969
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3969 Supplier OriGene Supplier No. CF811047
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PX19 (PRELID1) Monoclonal antibody specifically detects PX19 (PRELID1) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PX19 (PRELID1)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A6
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene GDI1
Gene Accession No. P31150
Gene Alias 25 kDa protein of relevant evolutionary and lymphoid interest; 2610524G07Rik; CGI-106; Preli; PRELI domain containing 1; PRELI domain-containing protein 1, mitochondrial; PRELID1; protein of relevant evolutionary and lymphoid interest; PX19; PX19 homolog; px19-like protein; RGD1308082; SBBI12
Host Species Mouse
Immunogen Full length human recombinant protein of human GDI1 produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2664
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.