Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PX19 (PRELID1) Monoclonal Antibody (OTI2B11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF810974


Description
PX19 (PRELID1) Monoclonal antibody specifically detects PX19 (PRELID1) in Human, Mouse samples. It is validated for Western BlotSpecifications
PX19 (PRELID1) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P13378, P23463 | |
Mouse | |
Protein A/G | |
RUO | |
15437, 3234 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI2B11 | |
Unconjugated | |
HOXD8 | |
25 kDa protein of relevant evolutionary and lymphoid interest; 2610524G07Rik; CGI-106; Preli; PRELI domain containing 1; PRELI domain-containing protein 1, mitochondrial; PRELID1; protein of relevant evolutionary and lymphoid interest; PX19; PX19 homolog; px19-like protein; RGD1308082; SBBI12 | |
Human recombinant protein fragment corresponding to amino acids 69-290 of human HOXD8 (NP_062458) produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction