Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PXDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564977
Description
Immunogen sequence: ISEIMRSNGFC LANTETIVID HSIPNGRDQQ LGVDPTEHLF ENGSEFPSEL EDGDDPAAYV TNLSYYHLVP FETDIWD Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
PXDC1 gene ontology annotations related to this gene include phosphatidylinositol binding.
Specifications
PXDC1 | |
Polyclonal | |
Unconjugated | |
PXDC1 | |
1300014I06Rik; C6orf145; PX domain containing 1; PX domain-containing protein 1; PX domain-containing protein C6orf145; PX domain-containing protein C6orf145 homolog; Pxdc1; RGD1311307 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221749 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TGL8 | |
PXDC1 | |
Recombinant Human PXDC1. Recombinant protein control fragment (Product #RP-90830). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction