Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PXDN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | PXDN |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121659
|
Invitrogen™
PA5121659 |
20 μL |
Each for $237.00
|
|
|||||
PIPA5121658
|
Invitrogen™
PA5121658 |
100 μL |
Each for $539.50
|
|
|||||
Description
Drosophila peroxidasin is an extracellular matrix-associated peroxidase (Horikoshi et al., 1999 [PubMed 10441517]). It is expressed exclusively in hemocytes derived from head mesoderm at a very early stage of differentiation. Peroxidasin exists as a homotrimer with a unique hybrid structure that combines an enzymatically functional peroxidase domain with motifs that are typically found in extracellular matrix-associated proteins. It is a secreted protein that contains a secretory recognition sequence at its N terminus. Peroxidasin catalyzes hydrogen peroxide-driven radioiodination, oxidations, and the formation of dityrosine in vitro. It is also thought to function in extracellular matrix consolidation, phagocytosis, and defense.Specifications
PXDN | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q92626 | |
7837 | |
Glu76-Gly260 (Accession Q92626), with N-terminal His Tag. | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
PXDN |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
2310075M15Rik; C85409; COPOA; D2S448; D2S448E; E330004E07; Kiaa0230; Melanoma-associated antigen MG50; MG50; mKIAA0230; p53-responsive gene 2 protein; PE responsive protein c64; Perc64; peroxidasin; peroxidasin homolog; peroxidasin homolog (Drosophila); PRG2; PXDN; PXN; Vascular peroxidase 1; VPO; VPO1 | |
PXDN | |
IgG | |
Antigen affinity chromatography, Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title