Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PXK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554970 Supplier Invitrogen™ Supplier No. PA554970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSLPLPPKKL IGNMDREFIA ERQKGLQNYL NVITTNHILS NCELVKKFLD PNNYSANYTE IALQQVSMFF RSEPKWEVVE PLKDIGWRIR KKYFLMKIKN QPKERLVLSW ADLGPDKYLS DKDFQCLIKL LP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

PXK is widely expressed in all tissues examined except in heart. It binds to and modulates brain Na,K-ATPase subunits ATP1B1 and ATP1B3 and may thereby participate in the regulation of electrical excitability and synaptic transmission. However, the protein kinase domain is predicted to be catalytically inactive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PXK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PXK
Gene Accession No. Q7Z7A4
Gene Alias C230080L11Rik; D14Ertd813e; EMBL:AAP92800.1, ECO:0000312; modulator of Na,K-ATPase; modulator of Na,K-ATPase long form; MONaKA; PX domain containing serine/threonine kinase; PX domain containing serine/threonine kinase like; PX domain-containing protein kinase-like protein; PX ser/thr kinase v2; PX serine/threonine kinase; pxk; pxk {ECO:0000312; RGD:727819}
Gene Symbols PXK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PXK. Recombinant protein control fragment (Product #RP-91475).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.