Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PXK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598562
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598562 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598562 Supplier Invitrogen™ Supplier No. PA598562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PXK Polyclonal Antibody for Western Blot, IHC (P), ELISA

PXK is widely expressed in all tissues examined except in heart. It binds to and modulates brain Na,K-ATPase subunits ATP1B1 and ATP1B3 and may thereby participate in the regulation of electrical excitability and synaptic transmission. However, the protein kinase domain is predicted to be catalytically inactive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PXK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PXK
Gene Accession No. Q4FZZ1, Q7Z7A4, Q8BX57
Gene Alias C230080L11Rik; D14Ertd813e; EMBL:AAP92800.1, ECO:0000312; modulator of Na,K-ATPase; modulator of Na,K-ATPase long form; MONaKA; PX domain containing serine/threonine kinase; PX domain containing serine/threonine kinase like; PX domain-containing protein kinase-like protein; PX ser/thr kinase v2; PX serine/threonine kinase; pxk; pxk {ECO:0000312; RGD:727819}
Gene Symbols PXK
Host Species Rabbit
Immunogen Recombinant Human PX domain-containing protein kinase-like protein (1-200AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218699, 306203, 54899
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.