Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PXMP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575898 Supplier Invitrogen™ Supplier No. PA575898
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Peroxisomes are subcellular organelles with important functions in lipid metabolism that are found in virtually all eucaryotic cells. The peroxisomal membrane contains a number of integral and peripheral membrane proteins involved in the import of peroxisomal matrix proteins and the transport of metabolites across the membrane. Pxmp2 (Peroxisomal membrane protein 2), also known as PMP22, is a 195 amino acid multi-pass membrane protein that belongs to the peroxisomal membrane protein PXMP2/4 family. The Pxmp2 protein interacts with both Peroxin 19 and Siva proteins. The Peroxin 19 protein binds multiple peroxisomal membrane proteins (PMPs), is predominantly cytoplasmic, and is required for peroxisome membrane synthesis. Pxmp2 is the most abundant peroxisomal membrane protein in higher eukaryotes. Pxmp2's expression is tissue-specific with highest levels of expression in liver, kidney and heart tissue. It has been suggested that Pxmp2 is involved in pore-forming activity and may contribute to the broad permeability of the peroxisomal membrane. Disruption of the mouse Pxmp2 gene leads to partial restriction of peroxisomal membrane permeability to solutes in vitro and in vivo. The Pxmp2 gene is conserved in chimpanzee, dog, cow, mouse, rat, zebrafish, fruit fly and mosquito, and maps to human chromosome 12q24.33.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PXMP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PXMP2
Gene Accession No. Q07066, Q9NR77
Gene Alias 22 kDa peroxisomal membrane protein; 22kDa; liver regeneration-related protein LRRG01; peroxisomal membrane protein 2; peroxisomal membrane protein 2, 22 kDa; peroxisomal membrane protein 2, 22kDa; PMP22; Pxmp2; T44
Gene Symbols PXMP2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human PXMP2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29533, 5827
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.