Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PXMP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114248 Supplier Invitrogen™ Supplier No. PA5114248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VWGLVLWLFE YHRSTLQPSL QSSMTYLYED SNVWHDISDF LVYNKSRPSN For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 85%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 75%; Rabbit: 92%; Rat: 83%.

PXMP4 belongs to the peroxisomal membrane protein PXMP2/4 family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PXMP4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PXMP4
Gene Accession No. Q9Y6I8
Gene Alias 24 kDa intrinsic membrane protein; 24 kDa peroxisomal intrinsic membrane protein; 3010018P03Rik; peroxisomal membrane protein 4; peroxisomal membrane protein 4, 24kDa; PMP24; PXMP4
Gene Symbols PXMP4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human PXMP4 (aa 163-212).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11264
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.