Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PXMP4 Antibody (7S41), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP221987C
Description
PXMP4 Monoclonal specifically detects PXMP4 in Mouse samples. It is validated for ELISA.Specifications
PXMP4 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PXMP4 | |
Mouse recombinant PXMP4 | |
0.1 mL | |
11264 | |
Store at 4C in the dark. | |
IgG2 |
ELISA | |
7S41 | |
ELISA | |
PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa | |
Rat | |
Protein G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction