Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PXR/NR1I2 Antibody (1D12G1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
| Antigen | PXR/NR1I2 |
|---|---|
| Clone | 1D12G1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61816
![]() |
Novus Biologicals
NBP261816 |
0.1 mL |
Each of 1 for $560.50
|
|
|||||
Description
PXR/NR1I2 Monoclonal specifically detects PXR/NR1I2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| PXR/NR1I2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR | |
| NR1I2 | |
| IgG2b | |
| Protein G purified |
| 1D12G1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| Stem Cell Markers, Transcription Factors and Regulators | |
| 8856 | |
| Purified recombinant fragment of human NR1I2 (AA: 1-142) expressed in E. Coli. | |
| Primary | |
| 9.8 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title