Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PXR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110975
Description
Immunogen sequence: VQGLTEEQRM MIRELMDAQM KTFDTTFSHF KNFRLPGVLS SGCELPESLQ APSREEAAKW SQVRKDLCSL KVSLQLRGED GSVWNYKPPA DSGGKEIFSL LPHMADMSTY MFKGIISFAK VISYFRDLPI EDQISLLK Highest antigen sequence indentity to the following orthologs: Rat - 72%, Mouse - 75%.
This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized.
Specifications
| PXR | |
| Polyclonal | |
| Unconjugated | |
| NR1I2 | |
| BXR; mPXR; Nr1i2; Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group 1, member 2; nuclear receptor subfamily 1, group I, member 2; ONR 1; ONR1; orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; PAR; PAR 1; PAR 2; PAR q; PAR1; PAR2; PARq; pregnane X nuclear receptor variant 2; Pregnane X receptor; pregnane X receptor (nuclear receptor sub family 1, group I, member 2); PRR; PXR; PXR.1; PXR.2; PXR1; SAR; Steroid and xenobiotic receptor; SXR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8856 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75469 | |
| NR1I2 | |
| Recombinant Protein corresonding to Human PXR. Recombinant protein control fragment (Product #RP-105811). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction