Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PXT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570123 Supplier Invitrogen™ Supplier No. PA570123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Horse: 92%; Human: 100%.

PXT1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PXT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PXT1
Gene Accession No. Q8NFP0
Gene Alias peroxisomal testis enriched protein 1; peroxisomal testis-specific protein 1; peroxisomal, testis specific 1; PXT1; Small testis-specific peroxisomal protein; STEPP
Gene Symbols PXT1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human PXT1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222659
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.