Learn More
Invitrogen™ PYCARD Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
Specifications
Specifications
| Antigen | PYCARD |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | PYCARD |
| Gene Accession No. | Q9EPB4, Q9ULZ3 |
| Gene Alias | 9130417A21Rik; Apoptosis-associated speck-like protein containing a CARD; Asc; CARD5; caspase recruitment domain-containing protein 5; hASC; mASC; MGC10332; Pycard; PYD and CARD domain containing; PYD and CARD domain-containing protein; Target of methylation-induced silencing 1; TMS; TMS1; TMS-1; TNS1 |
| Gene Symbols | PYCARD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.