Learn More
PYCR1 Mouse anti-Human, Clone: OTI4F2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811756

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline. This enzyme may also play a physiologic role in the generation of NADP(+) in some cell types. The protein forms a homopolymer and localizes to the mitochondrion. Alternate splicing results in two transcript variants encoding different isoforms.Specifications
PYCR1 | |
Monoclonal | |
Unconjugated | |
PYCR1 | |
ARCL2B, ARCL3B, P5C, P5CR, PIG45, PP222, PRO3, PYCR | |
Mouse | |
Affinity Chromatography | |
RUO | |
5831 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4F2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P32322 | |
PYCR1 | |
Human recombit protein fragment corresponding to amino acids 1-319 of human PYCR1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.