Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYCR1 Mouse anti-Human, Mouse, Rat, Clone: 1H2G1, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 665101IG150UL
Description
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline. This enzyme may also play a physiologic role in the generation of NADP(+) in some cell types. The protein forms a homopolymer and localizes to the mitochondrion. Alternate splicing results in two transcript variants encoding different isoforms.Specifications
| PYCR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P32322, Q922W5 | |
| Pycr1 | |
| PYCR1 Fusion Protein Ag17919 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1H2G1 | |
| Unconjugated | |
| PYCR1 | |
| P5C, P5C reductase 1, P5CR, P5CR 1, PIG45, PP222, PYCR, PYCR1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 209027, 287877, 5831 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction