Learn More
PYCR1/P5CR Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304837AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline. This enzyme may also play a physiologic role in the generation of NADP(+) in some cell types. The protein forms a homopolymer and localizes to the mitochondrion. Alternate splicing results in two transcript variants encoding different isoforms.Specifications
PYCR1/P5CR | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P32322 | |
PYCR1 | |
Between 269 and 319 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PYCR1 | |
ARCL2B, ARCL3B, P5C, P5CR, PIG45, PP222, PRO3, PYCR, P5C reductase 1, P5CR 1, mitochondrial pyrroline-5-carboxylate reductase 1, proliferation-inducing protein 45, pyrroline-5-carboxylate reductase 1, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5831 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.