Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PYCR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599986
Description
Antibody detects endogenous levels of total PYCR1.
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline. This enzyme may also play a physiologic role in the generation of NADP(+) in some cell types. The protein forms a homopolymer and localizes to the mitochondrion. Alternate splicing results in two transcript variants encoding different isoforms.
Specifications
PYCR1 | |
Polyclonal | |
Unconjugated | |
PYCR1 | |
ARCL2B; ARCL3B; P5C; P5C reductase 1; P5CR; P5CR 1; PIG45; PP222; PRO3; proliferation-inducing protein 45; PYCR; Pycr1; pyrroline-5-carboxylate reductase 1; pyrroline-5-carboxylate reductase 1, mitochondrial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
209027, 287877, 5831 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P32322, Q922W5 | |
PYCR1 | |
A synthesized peptide derived from human PYCR1(Accession P32322), corresponding to amino acid residues Q151-L201. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction