Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYCR2 Antibody (OTI3D1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273743IR
Description
PYCR2 Monoclonal specifically detects PYCR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PYCR2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PYCR2 | |
Full length recombinant protein of human PYCR2 (NP_037460) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3D1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 1.5.1.2, FLJ54750, P5C reductase 2, P5CR 2, P5CR2, pyrroline 5-carboxylate reductase isoform, pyrroline-5-carboxylate reductase 2, pyrroline-5-carboxylate reductase family, member 2 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
29920.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction