Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PYCR2 Monoclonal Antibody (OTI2A12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525282
Description
PYCR2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
PYCR2 belongs to the pyrroline-5-carboxylate reductase family. The function of the PYCR2 protein is not known.
Specifications
PYCR2 | |
Monoclonal | |
0.86 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96C36 | |
Pycr2 | |
Full length human recombinant protein of PYCR2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI2A12 | |
Unconjugated | |
Pycr2 | |
1810018M05Rik; HLD10; P5C reductase 2; P5CR 2; P5CR2; Pycr2; pyrroline 5-carboxylate reductase isoform; pyrroline-5-carboxylate reductase 2; pyrroline-5-carboxylate reductase family member 2; pyrroline-5-carboxylate reductase family, member 2; QtsA-16987; unnamed protein product | |
Mouse | |
Affinity Chromatography | |
RUO | |
29920 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction