Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYCR2 Monoclonal Antibody (OTI4A10)
GREENER_CHOICE

Catalog No. PIMA525277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525277 Supplier Invitrogen™ Supplier No. MA525277
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PYCR2 Monoclonal Antibody for Western Blot, ICC/IF, Flow

PYCR2 belongs to the pyrroline-5-carboxylate reductase family. The function of the PYCR2 protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYCR2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4A10
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pycr2
Gene Accession No. Q96C36
Gene Alias 1810018M05Rik; HLD10; P5C reductase 2; P5CR 2; P5CR2; Pycr2; pyrroline 5-carboxylate reductase isoform; pyrroline-5-carboxylate reductase 2; pyrroline-5-carboxylate reductase family member 2; pyrroline-5-carboxylate reductase family, member 2; QtsA-16987; unnamed protein product
Gene Symbols Pycr2
Host Species Mouse
Immunogen Full length human recombinant protein of PYCR2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29920
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.