Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYCRL Antibody (OTI1B12), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273752B
Description
PYCRL Monoclonal specifically detects PYCRL in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PYCRL | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PYCR3 | |
Full length recombinant protein of human PYCRL (NP_075566) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1B12 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 1.5.1.2, FLJ13852, P5C reductase 3, P5CR 3, pyrroline-5-carboxylate reductase 3, pyrroline-5-carboxylate reductase-like, Pyrroline-5-carboxylate reductase-like protein | |
Mouse | |
Affinity Purified | |
metabolism | |
65263.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction