Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYCRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110247 Supplier Invitrogen™ Supplier No. PA5110247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAEPSPRR VGFVGAGRMA GAIAQGLIRA GKVEAQHILA SAPTDRNLCH FQALGCRTTH SNQEVLQSCL LVIFATKPHV LPAVLAEVAP VVTTEHILVS VAAGVSLSTL EELLPPNTRV LRVLPNLPCV VQEGAIVMAR GRHVGSSETK LLQHLLEACG RCEEVPEAYV DIHTGLSGSG VAFVCAFSEA LAEGAVKMGM PSSLAHRIAA QTLLGTAKML LHEGQHPAQL RSDVCTPGGT TIYGLHALEQ GGLRAATMSA VEAATCRAKE LSRK.

PYCRL encodes a protein that belongs to the pyrroline-5-carboxylate reductase family of enzymes. Members of this family catalyze the final step in proline biosynthesis, converting pyrroline-5-carboxylate to proline. Glutamate and ornithine are precursors in the synthesis of proline. The protein encoded by this gene is a cytoplasmic enzyme involved in the biosynthesis of proline from ornithine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYCRL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pycrl
Gene Accession No. Q9DCC4
Gene Alias 1110058B13Rik; 2700073G24Rik; P5C reductase 3; P5CR 3; PYCR3; Pycrl; pyrroline-5-carboxylate reductase 3; pyrroline-5-carboxylate reductase-like; pyrroline-5-carboxylate reductase-like protein
Gene Symbols Pycrl
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-274 of human PYCRL (NP_075566.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66194
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.