Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYDC1 Mouse anti-Human, Clone: OTI11C3, lyophilized, TrueMAB™
SDP

Catalog No. 501666567
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-6567 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-6567 Supplier OriGene Supplier No. CF803556
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

ASC2 associates with PYCARD/ASC and modulates its ability to collaborate with MEFV/pyrin and NLRP3/cryopyrin in NF-kappa-B and pro-caspase-1 activation. It suppresses kinase activity of NF-kappa-B inhibitor kinase (IKK) complex, expression of NF-kappa-B inducible genes and inhibits NF-kappa-B activation by cytokines and LPS.

Specifications

Antigen PYDC1
Applications Western Blot
Classification Monoclonal
Clone OTI11C3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PYDC1
Gene Accession No. Q8WXC3
Gene Alias ASC2, POP1, PYC1, cPOP1
Gene Symbols PYDC1
Host Species Mouse
Immunogen Full length human recombit protein of human PYDC1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260434
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.