Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102846
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102846 Supplier Invitrogen™ Supplier No. PA5102846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ASC2.

ASC2 associates with PYCARD/ASC and modulates its ability to collaborate with MEFV/pyrin and NLRP3/cryopyrin in NF-kappa-B and pro-caspase-1 activation. It suppresses kinase activity of NF-kappa-B inhibitor kinase (IKK) complex, expression of NF-kappa-B inducible genes and inhibits NF-kappa-B activation by cytokines and LPS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYDC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PYDC1
Gene Accession No. Q8WXC3
Gene Alias ASC2; ASCI; cellular POP1; cPOP1; PAAD-only protein 1; POP1; PYC1; PYD (pyrin domain) containing 1; PYDC1; pyrin domain containing 1; pyrin domain-containing protein 1; Pyrin-only protein 1
Gene Symbols PYDC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PYDC1(Accession Q8WXC3), corresponding to amino acid residues M1-P31.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260434
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.