Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYGB Polyclonal Antibody

Catalog No. PIPA5121953
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121953 20 μL
PIPA5119152 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121953 Supplier Invitrogen™ Supplier No. PA5121953
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PYGB Polyclonal Antibody for IHC (P), Western Blot

The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYGB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PYGB
Gene Accession No. P11216, Q8CI94
Gene Alias brain glycogen phosphorylase; glycogen phosphorylase B; glycogen phosphorylase, brain form; GLYPHOA; GPBB; MGC9213; phosphorylase, glycogen; brain; PYGB
Gene Symbols PYGB
Host Species Rabbit
Immunogen Recombinant protein Glycogen Phosphorylase, Brain. The antigen corresponds to amino acid range 477-731 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110078, 5834
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.