Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYGL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552054 Supplier Invitrogen™ Supplier No. PA552054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVAATLQDII RRFKASKFGS TRGAGTVFDA FPDQVAIQLN DTHPALAIPE LMRIFVDIEK LPWSKAWELT QKTFAYTNHT VLPEALERWP VDLVEKLLPR HLEIIYEINQ KHLDRIVALF PKDVDRLRRM SLI Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

This gene encodes a homodimeric protein that catalyses the cleavage of alpha-1,4-glucosidic bonds to release glucose-1-phosphate from liver glycogen stores. This protein switches from inactive phosphorylase B to active phosphorylase A by phosphorylation of serine residue 15. Activity of this enzyme is further regulated by multiple allosteric effectors and hormonal controls. Humans have three glycogen phosphorylase genes that encode distinct isozymes that are primarily expressed in liver, brain and muscle, respectively. The liver isozyme serves the glycemic demands of the body in general while the brain and muscle isozymes supply just those tissues. In glycogen storage disease type VI, also known as Hers disease, mutations in liver glycogen phosphorylase inhibit the conversion of glycogen to glucose and results in moderate hypoglycemia, mild ketosis, growth retardation and hepatomegaly. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYGL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PYGL
Gene Accession No. P06737
Gene Alias glycogen liver; Glycogen phosphorylase, liver form; GPLL; GSD6; Lgp; liver glycogen phosphorylase; phosphorylase, glycogen, liver; PYGL
Gene Symbols PYGL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PYGL. Recombinant protein control fragment (Product #RP-88882).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5836
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.