Learn More
Invitrogen™ PYGL Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Dog (100%); Gibbon, Marmoset, Panda (90%); Mouse, Sheep, Hamster, Horse, Rabbit (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Specifications
Specifications
| Antigen | PYGL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | PYGL |
| Gene Accession No. | P06737 |
| Gene Alias | glycogen liver; Glycogen phosphorylase, liver form; GPLL; GSD6; Lgp; liver glycogen phosphorylase; phosphorylase, glycogen, liver; PYGL |
| Gene Symbols | PYGL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.