Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYGM Mouse anti-Human, Mouse, Rat, Clone: OTI5D1, liquid, TrueMAB™
SDP

Catalog No. 501683965
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3965 Supplier OriGene Supplier No. TA811300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PYGM encodes a muscle enzyme involved in glycogenolysis. Highly similar enzymes encoded by different genes are found in liver and brain. Mutations in this gene are associated with McArdle disease (myophosphorylase deficiency), a glycogen storage disease of muscle. Alternative splicing results in multiple transcript variants.

Specifications

Antigen PYGM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PYGM
Gene Accession No. P09812, P11217, Q9WUB3
Gene Alias glycogen phosphorylase, muscle associated
Gene Symbols PYGM
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 698-842 of human PYGM produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19309, 24701, 5837
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.