Learn More
Invitrogen™ PYGM Polyclonal Antibody

Description
Positive test controls include: Mouse skeletal muscle, Mouse brain, Mouse lung, Rat skeletal muscle. Immunogen sequence: IEQLSSGFFS PKQPDLFKDI VNMLMHHDRF KVFADYEDYI KCQEKVSALY KNPREWTRMV IRNIATSGKF SSDRTIAQYA REIWGVEPSR QRLPAPDEAI.
Specifications
Specifications
| Antigen | PYGM |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.86 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | PYGM |
| Gene Accession No. | P09812, P11217, Q9WUB3 |
| Gene Alias | AI115133; glycogen muscle; glycogen phosphorylase; glycogen phosphorylase, muscle associated; glycogen phosphorylase, muscle form; GPMM; muscle (McArdle syndrome); muscle glycogen phosphorylase; Muscpho; Myophosphorylase; PG; Phosphorylase glycogen; phosphorylase, glycogen, muscle; Phosphorylase, glycogen; muscle (McArdle syndrome); Pygm; unnamed protein product |
| Gene Symbols | PYGM |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.