Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PYGM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119155
This item is not returnable.
View return policy
Description
PYGM Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a muscle enzyme involved in glycogenolysis. Highly similar enzymes encoded by different genes are found in liver and brain. Mutations in this gene are associated with McArdle disease (myophosphorylase deficiency), a glycogen storage disease of muscle. Alternative splicing results in multiple transcript variants.
Specifications
PYGM | |
Polyclonal | |
Unconjugated | |
PYGM | |
AI115133; glycogen muscle; glycogen phosphorylase; glycogen phosphorylase, muscle associated; glycogen phosphorylase, muscle form; GPMM; muscle (McArdle syndrome); muscle glycogen phosphorylase; Muscpho; Myophosphorylase; PG; Phosphorylase glycogen; phosphorylase, glycogen, muscle; Phosphorylase, glycogen; muscle (McArdle syndrome); Pygm; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
19309, 24701, 5837 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P09812, P11217, Q9WUB3 | |
PYGM | |
Recombinant protein Glycogen Phosphorylase, Muscle. The antigen corresponds to amino acid range 11-187 of the target protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction