Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYGO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547501
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547501 Supplier Invitrogen™ Supplier No. PA547501
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Pygopus 1 (PYGO1) is a gene located on chromosome 6p21.31, encoding a protein that plays a crucial role in the Wnt signaling pathway, which is fundamental for various developmental processes including cell proliferation, polarity, and fate determination. PYGO1 is a part of the Pygopus family of proteins, characterized by their Plant Homeodomain (PHD) finger domain that facilitates interaction with histones. Within the Wnt signaling pathway, PYGO1 acts as a coactivator by binding to components of the transcription complex, thus aiding in the expression of Wnt target genes. The protein is especially important in embryonic development and is implicated in specific processes such as limb formation and neural development. Dysregulation of PYGO1 and the Wnt pathway is associated with developmental disorders and certain cancers, demonstrating the protein's significance in maintaining cellular homeostasis. Current studies are focusing on the mechanistic aspects of PYGO1's function in transcription regulation and its potential role in therapeutic strategies targeting Wnt signaling-related diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYGO1
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PYGO1
Gene Accession No. Q9D0P5
Gene Alias 2600014C22Rik; PYGO1; pygopus 1; pygopus family PHD finger 1; pygopus homolog 1; pygopus-like protein 1; zgc:172214
Gene Symbols PYGO1
Host Species Goat
Immunogen E. coli-derived recombinant mouse Pygopus-1 Ser2-Ala417.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72135
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.