Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYGO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559927 Supplier Invitrogen™ Supplier No. PA559927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RMPPHVPPRM SSPYCGPYSL RNQPHPFPQN PLGMGFNRPH AFNFGPHDNS SFGNPSYNNA LSQNVNMPNQ HFRQNPAENF SQIPPQNASQ VSNPDLASNF VPGNNSNFTS PLESNHSFIP PPNTFGQAKA PPPKQDFTQG ATK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

Pygopus 1 (PYGO1) is a gene located on chromosome 6p21.31, encoding a protein that plays a crucial role in the Wnt signaling pathway, which is fundamental for various developmental processes including cell proliferation, polarity, and fate determination. PYGO1 is a part of the Pygopus family of proteins, characterized by their Plant Homeodomain (PHD) finger domain that facilitates interaction with histones. Within the Wnt signaling pathway, PYGO1 acts as a coactivator by binding to components of the transcription complex, thus aiding in the expression of Wnt target genes. The protein is especially important in embryonic development and is implicated in specific processes such as limb formation and neural development. Dysregulation of PYGO1 and the Wnt pathway is associated with developmental disorders and certain cancers, demonstrating the protein's significance in maintaining cellular homeostasis. Current studies are focusing on the mechanistic aspects of PYGO1's function in transcription regulation and its potential role in therapeutic strategies targeting Wnt signaling-related diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYGO1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PYGO1
Gene Accession No. Q9Y3Y4
Gene Alias 2600014C22Rik; PYGO1; pygopus 1; pygopus family PHD finger 1; pygopus homolog 1; pygopus-like protein 1; zgc:172214
Gene Symbols PYGO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PYGO1. Recombinant protein control fragment (Product #RP-89401).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.