Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pygopus-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321376100UL
Description
Pygopus-2 Polyclonal antibody specifically detects Pygopus-2 in Human samples. It is validated for ImmunofluorescenceSpecifications
Pygopus-2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
1190004M21Rik, FLJ33226, pygopus 2, pygopus homolog 2, pygopus homolog 2 (Drosophila) | |
This antibody was developed against Recombinant Protein corresponding to amino acids: PSTGRKQGKAGLQMKSPEKKRRKSNTQGPAYSHLTEFAPPPTPMVDHLVASNPFEDDFGAPKVGVAAPPF | |
100 μg | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
90780 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction