Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYHIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562350 Supplier Invitrogen™ Supplier No. PA562350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IESIPVKGII PSKKTKQKEV YPATPACTPS NRLTAKGAEE TLGPQKRKKP SEEETGTKRS KMSKEQTRPS CS Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 30%.

PYHIN1 belongs to the HIN200 family of interferon-inducible proteins that share a 200-amino acid signature motif at their C-terminal ends. HIN200 proteins are primarily nuclear and are involved in transcriptional regulation of genes important for cell cycle control, differentiation, and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYHIN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PYHIN1
Gene Accession No. Q6K0P9
Gene Alias IFIX; interferon-inducible protein X; PYHIN1; pyrin and HIN domain family member 1; pyrin and HIN domain family, member 1; pyrin and HIN domain-containing protein 1; RP11-520H16.1
Gene Symbols PYHIN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PYHIN1. Recombinant protein control fragment (Product #RP-101415).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149628
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.