Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYHIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525293
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525293 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525293 Supplier Invitrogen™ Supplier No. PA525293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PYHIN1 Polyclonal Antibody for Western Blot, IHC (P), Flow

PYHIN1 belongs to the HIN200 family of interferon-inducible proteins that share a 200-amino acid signature motif at their C-terminal ends. HIN200 proteins are primarily nuclear and are involved in transcriptional regulation of genes important for cell cycle control, differentiation, and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYHIN1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PYHIN1
Gene Accession No. Q6K0P9
Gene Alias IFIX; interferon-inducible protein X; PYHIN1; pyrin and HIN domain family member 1; pyrin and HIN domain family, member 1; pyrin and HIN domain-containing protein 1; RP11-520H16.1
Gene Symbols PYHIN1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 26-55 amino acids from the N-terminal region of human PYHIN1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149628
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.