Learn More
Promega PYK2 Kinase Enzyme System, 1mg
Catalog No. PRV6477 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6477 1 mg
1 options
Catalog No. PRV6477 Supplier Promega Supplier No. V6477
Add to Cart
Add to Cart
Request a Quote
Recombinant human PYK2 (360-690) was expressed by baculovirus in Sf9 cells using an N-terminal His tag. PYK2 (also known as FAK2/RAFTK) is a member of the focal adhesion PTK family.
  • Easily Screen and Profile PYK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg