Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115810 Supplier Invitrogen™ Supplier No. PA5115810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PYK2.

PTK2B (FAK2) is a cytoplasmic tyrosine kinase involved in calcium-induced regulation of ion channels and activation of the map kinase signaling pathway. PTK2B regulates reorganization of the actin cytoskeleton, cell polarization, cell migration, adhesion, spreading, and bone remodeling. FAK2 plays a role in the regulation of the humoral immune response and is required for normal levels of marginal B-cells in the spleen and normal migration of splenic B-cells. It functions in signaling downstream of integrin and collagen receptors, immune receptors, G-protein coupled receptors, cytokine, chemokine, and growth factor receptors, and mediates responses to cellular stress. Aberrant FAK2 expression may play a role in cancer cell proliferation, migration, and invasion in tumor formation and metastasis. Elevated expression has been seen in gliomas, hepatocellular carcinoma, lung cancer, and breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYK2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PTK2B
Gene Accession No. P70600, Q14289, Q9QVP9
Gene Alias CADTK; CAK beta; CAKB; CAKbeta; CAK-beta; calcium-dependent tyrosine kinase; Calcium-regulated non-receptor proline-rich tyrosine kinase; cell adhesion kinase beta; cellular adhesion kinase beta; E430023O05Rik; FADK; FADK 2; FADK2; FAK; FAK1; FAK2; focal adhesion kinase 2; FRNK; PKB; pp125FAK; proline-rich tyrosine kinase 2; protein kinase B; protein tyrosine kinase 2 beta; protein-tyrosine kinase 2-beta; PTK; PTK2 protein tyrosine kinase 2 beta; PTK2B; PTK2B protein tyrosine kinase 2 beta; PYK2; RAFTK; related adhesion focal tyrosine kinase
Gene Symbols PTK2B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PTK2B(Accession Q14289), corresponding to amino acid residues L376-V426.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19229, 2185, 50646
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.