Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYK2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501567179
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7179 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7179 Supplier Bethyl Laboratories, Inc Supplier No. A304226AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Focal adhesion kinase subfamily consist of the non-receptor proline-rich protein tyrosine kinases (PKTs). Two members of the family are focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). These two kinases have molecular mass between 110-125 kDa and are closely related in their structure. The presence of two proline-rich motifs within the C-terminal domains is conserved.

Specifications

Antigen PYK2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PTK2B
Gene Accession No. Q14289
Gene Alias FADK, FAK, FAK1, FRNK, pp125FAK
Gene Symbols PTK2B
Host Species Rabbit
Immunogen Between 275 and 325
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2185
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.