Learn More
PYK2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304227AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
Focal adhesion kinase subfamily consist of the non-receptor proline-rich protein tyrosine kinases (PKTs). Two members of the family are focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). These two kinases have molecular mass between 110-125 kDa and are closely related in their structure. The presence of two proline-rich motifs within the C-terminal domains is conserved.Specifications
PYK2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q14289 | |
PTK2B | |
Between 959 and 1009 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PTK2B | |
FADK, FAK, FAK1, FRNK, pp125FAK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2185 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.