Learn More
PYK2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304226A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Focal adhesion kinase subfamily consist of the non-receptor proline-rich protein tyrosine kinases (PKTs). Two members of the family are focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). These two kinases have molecular mass between 110-125 kDa and are closely related in their structure. The presence of two proline-rich motifs within the C-terminal domains is conserved.Specifications
PYK2 | |
Polyclonal | |
Unconjugated | |
PTK2B | |
FADK, FAK, FAK1, FRNK, pp125FAK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2185 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q14289 | |
PTK2B | |
Between 275 and 325 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.