Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pyr 10
GREENER_CHOICE

Catalog No. 694110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-411-0 10 mg
69-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-411-0 Supplier Tocris Bioscience™ Supplier No. 6941/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective TRPC3 inhibitor

Pyr 10 is a TRPC3 (transient receptor potential canonical 3) channel inhibitor (IC50 = 0.72 μM for TRPC3 mediated calcium entry in vitro), which displays approximately 18-fold selectivity for TRPC3-mediated receptor operated calcium entry (ROCE) over STIM1/Orai1-mediated store operated calcium entry (SOCE). Pyr 10 inhibits NFATc3 activation and inhibits proliferation of rat ventricular cardiac fibroblasts.

Specifications

Chemical Name or Material N-[4-[3,5-bis(Trifluoromethyl)-1 H-pyrazol-1-yl]phenyl]-4-methylbenzenesulfonamide
CAS 1315323-00-2
Quantity 10 mg
Target TRPC Channel Blockers
Molecular Formula C18H13F6N3O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.